Anti-VPREB3 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB09713
EVBIEB09713EA 202 EUR
EVBIEB09713
Anti-VPREB3 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: VPREB3 (pre-B lymphocyte 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR