Anti-TIMP1 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB11201
EVBIEB11201EA 339 EUR
EVBIEB11201
Anti-TIMP1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TIMP1 (TIMP metallopeptidase inhibitor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR