Anti-LRP1 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB12608
EVBIEB12608EA 200 EUR
EVBIEB12608
Anti-LRP1 Goat Polyclonal Antibody
Western Blot: Not yet tested - our routinely used western blotting protocol does not allow detection of proteins as large as the calculated size of 505kDa according to NP_002323.2. Therefore we cannot recommend an optimal concentration and the antibody i

Type: Primary
Antigen: LRP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Pig, Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR