Hand cleansing paste, Solopol® natural
Lieferant: Deb-STOKO
33456 33282
EVON33456EA 133 EUR
EVON33456 EVON33282
Hand cleansing paste, Solopol® natural
Hautpflege Seifen und Hautreiniger
Solvent-free hand cleanser paste with scrubbing-agent for extra heavy-duty contamination from, for example, oils, greases, carbon black, graphite, metallic dust, lubricants. For use in general industrial environments.
  • Astopon® natural scrubbing agent gently removes contaminants without damaging the skin
  • Contains skin protectant Eucornol® and aloe vera
  • Formulation leaves hands feeling smooth and conditioned
  • Solvent-free, pH neutral to the skin
Standards: EU Ecolabel
Bitte beachten *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904