Estesol® FX™ PURE Hand Cleansers
Lieferant: Deb-STOKO
EPU1L
EVONEPU1LEA 113 EUR
EVONEPU1L
Estesol® FX™ PURE Hand Cleansers
Hautpflege Seifen und Hautreiniger
Solvent-free PowerFOAM hand cleaner designed for professional use to remove industrial dust, dirt and food related soilings. Perfume-free and dye-free.
  • Quick and easy to use
  • Unique POWER FOAM formula
  • Ideal for high frequency use
  • Mild preservative
  • Formulated with a low pH value (4 to 5)
  • Effective, solvent free formula
  • Silicone-free and skin hypoallergenic
For use in general industrial and food manufacturing/processing environments to remove light duty oils, greases, general grime and food related soilings.
Standards: ECARF certified; EU Ecolabel certified.