Scissors, Medical Grade, Extra Fine Blades
Lieferant: EXCELTA
361
EXCE361EA 88.4 EUR
EXCE361
Scissors, Medical Grade, Extra Fine Blades
Scheren
These medical grade scissors are made of stainless steel.
  • Excellent for ultra-miniature work in confined areas
  • Cleanroom safe, medical grade stainless steel
  • Extremely precise cutting edges
  • Scissors, medical grade, 4 star quality
  • 17° curved
  • Stainless steel AISI 420
They are ideal for ultra miniature work in confined areas.