Exsil SAX, HPLC Columns
Lieferant: Avantor
EXSAX-5-250PEA 0 EUR
EXSAX-5-250P
Exsil SAX, HPLC Columns
Chromatographiesäulen
Exsil HPLC columns are a useful alternative to the classical spherical HPLC silicas.
  • Spherical porous silica
  • Competitively priced
  • High efficiency
Exsil 100, the standard material, is supplied in a range of particle sizes and bonded phases to cover a wide range of applications. Exsil 80 is a higher surface area, complementary range of materials.