Fast Blue BB base, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
Synonyme:
4'-Amino-2',5'-diethoxybenzanilide, N-(4-Amino-2,5-diethoxyphenyl)benzamide, Azoic Diazo No. 20, Fast blue BB, C.I. 37175, Azoic diazo component 20 (base)
F0125-25G
F0125-25GEA
141
EUR
F0125-25G
Fast Blue BB base, Sigma-Aldrich®
Fast Blue BB base
N-(4-Amino-2,5-diethoxyphenyl)benzamide, (4'-Amino-2',5'-diethoxybenzanilide; Azoic Diazo No. 20; Fast blue BB)
Formel:
C₁₇H₂₀N₂O₃ Molecular Weight: 300,36 g/mol Schmelzpunkt: 98…100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009091 CAS-Nummer: 120-00-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...