Fast Blue RR Salt, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Azoic Diazo No. 24, 4-Benzoylamino-2,5-dimethoxybenzenediazonium chloride hemi(zinc chloride) salt

F0500-25G
F0500-25GEA 551 EUR
F0500-25G
Fast Blue RR Salt, Sigma-Aldrich®
Fast Blue RR Salt

Fast blue RR salt has been used in the immunohistochemistry to detect the activity of alkaline phosphatase. It has been used in the procedure of connective tissue staining using picrosirius red.

Formel: C₁₅H₁₄ClN₃O₃
Molecular Weight: 387,89 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00074762
CAS-Nummer: 14726-29-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0023331de50580c174f15ca46f8f85a9955444eb&vid=f347cec7fefa44c1f7dbd77d4a9b7e845ce1aa53&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR