Fast Dark Blue R Salt, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
Azoic Diazo No. 51, Azoic Diazo Component 51
F0750-25G
F0750-25GEA
56.1
EUR
F0750-25G
Fast Dark Blue R Salt, Sigma-Aldrich®
Fast Dark Blue R Salt
Fast Dark Blue R Salt, (Azoic Diazo Component 51; Azoic Diazo No. 51)
Formel:
C₁₄HCl₂N₅O₄ Lagertemperatur: Gefrierschrank |
MDL:
MFCD00135879 CAS-Nummer: 6709-58-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...