FURA-2 pentapotassium salt, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
1-[2-(5-Carboxyoxazol-2-yl)-6-aminobenzofuran-5-oxy]-2-(2'-amino-5'-methyl-phenoxy)ethane-N,N,N',N'-tetraacetic acid . 5K, FURA-2
F0763-1MG
F0763-1MGEA
464
EUR
F0763-1MG
FURA-2 pentapotassium salt, Sigma-Aldrich®
FURA-2 pentapotassium salt
Cell impermeable fluorescent probe for Ca2+.
Formel:
C₂₉H₂₂K₅N₃O₁₄ Molecular Weight: 832 g/mol Lagertemperatur: Gefrierschrank |
CAS-Nummer:
113694-64-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...