Fura 2-AM ≥95% (durch HPLC)

Lieferant: SIGMA ALDRICH MICROSCOPY


M-Clarity™ Qualitätsstufe = MQ100

F0888-.5MG F0888-1MG
F0888-.5MGEA 381 EUR
F0888-.5MG F0888-1MG
Fura 2-AM ≥95% (durch HPLC)
Fura 2-AM

Fura 2-AM (Fura 2) is a calcium-specific ratio-metric fluorescent dye. It is also known as acetoxy-methyl-ester. In response to calcium ions, ratio metric dyes like Fura 2 changes their respective excitation or emission spectra. The intracellular calcium concentration is determined by the fluorescence emission ratio or excitation at distinct wavelengths. Fura 2 has an emission peak at 505 nm, and the excitation peak ranges between 340 to 380 nm and is the preferred calcium indicator.


  • Fura 2-AM is a popular probe used to determine dynamic changes in cytosolic free calcium in intact living cells


The major benefits of using the ester form of Fura 2 as Fura 2-AM are: It is cell-permeable. It can be loaded into cells easily. Another advantage of using Fura 2-AM over single wavelength dyes is that the ratio signal is independent of : The dye concentration. Illumination intensity, and Optical path length.

Formel: C₄₄H₄₇N₃O₂₄
Lagertemperatur: Gefrierschrank
MDL: MFCD00036976
CAS-Nummer: 108964-32-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f504ee9b37bbed37b388bd063880d5712589d44c&vid=a27a1e6f769056bce8e56ba9c7d5c94512b45dfc&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Yellow to Yellow-Green
Appearance (Form) Powder
Solubility (Colour) Yellow to Yellow-Green
Solubility (Turbidity)
10 mg/ml, acetone
Clear
Purity (HPLC) ≥95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR