Fura 2-AM ≥95% (durch HPLC)
Lieferant: SIGMA ALDRICH MICROSCOPY
M-Clarity™ Qualitätsstufe = MQ100
Fura 2-AM (Fura 2) is a calcium-specific ratio-metric fluorescent dye. It is also known as acetoxy-methyl-ester. In response to calcium ions, ratio metric dyes like Fura 2 changes their respective excitation or emission spectra. The intracellular calcium concentration is determined by the fluorescence emission ratio or excitation at distinct wavelengths. Fura 2 has an emission peak at 505 nm, and the excitation peak ranges between 340 to 380 nm and is the preferred calcium indicator.
- Fura 2-AM is a popular probe used to determine dynamic changes in cytosolic free calcium in intact living cells
The major benefits of using the ester form of Fura 2 as Fura 2-AM are: It is cell-permeable. It can be loaded into cells easily. Another advantage of using Fura 2-AM over single wavelength dyes is that the ratio signal is independent of : The dye concentration. Illumination intensity, and Optical path length.
Formel:
C₄₄H₄₇N₃O₂₄ Lagertemperatur: Gefrierschrank |
MDL:
MFCD00036976 CAS-Nummer: 108964-32-5 |
|
Spezifikation Testergebnisse
Appearance (Colour) | Yellow to Yellow-Green |
Appearance (Form) | Powder |
Solubility (Colour) | Yellow to Yellow-Green |
Solubility (Turbidity) 10 mg/ml, acetone |
Clear |
Purity (HPLC) | ≥95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...