Fast Red RC Salt, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

F2256-25G
F2256-25GEA 37 EUR
F2256-25G
Fast Red RC Salt, Sigma-Aldrich®
Fast Red RC Salt

Fast Red RC Salt, (2-Amino-4-chloroanisole diazotated zinc chloride complex; 5-Chloro-2-methoxybenzenediazonium chloride hemi(zinc chloride) salt; Diazo Fast Red RC)

Formel: C₇H₆Cl₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00074764
CAS-Nummer: 68025-25-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d7d5a5be1a95b52444c9b5b960c3ba6caf51a50&vid=f7426940f54fad756adfd772d62c2f0a29ae38e0&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR