Fast Red Violet LB Salt 90 Dye, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: 5-Chloro-4-benzamido-2-methylbenzenediazonium chloride hemi(zinc chloride) salt

MQ100

F3381-100MG F3381-1G F3381-250MG F3381-500MG F3381-5G
F3381-100MGEA 48.1 EUR
F3381-100MG F3381-1G F3381-250MG F3381-500MG F3381-5G
Fast Red Violet LB Salt 90 Dye, Sigma-Aldrich®
Fast Red Violet LB Salt

Fast Red Violet LB Salt has been used for the detection of tartrate-resistant acid phosphatase (TRAP) in tissue sections.


Fast Red Violet LB Salt, also known as 4′-amino-2′-chloro-5′-methylbenzanilide. It is a lipophilic amine with an aromatic system of moderate size and a hydrophilic diazonium cation. The diazonium salt is available commercially as a zinc chloride double salt. It comprises ≥90% dye content.

Formel: C₁₄H₁₁Cl₂N₃O
Molecular Weight: 376.31 g/mol
Lagertemperatur: Room Temperature
MDL: MFCD00011892
CAS-Nummer: 32348-81-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31080c577200ca6c8a052d50e030a944503fe229&vid=31080c577200ca6c8a052d50e030a944503fe229&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Carbon Content 40.2 - 49.1%
Nitrogen Content 10.1 - 12.3%
Dye Content ≥90%
Solubility (Turbidity), 1 mg/ml, Water Clear to Slightly Hazy
Solubility (Colour) Very Faint Yellow to Yellow


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR