Aqueous mounting medium, Fluoromount™, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
F4680-25ML
F4680-25MLEA
142
EUR
F4680-25ML
Aqueous mounting medium, Fluoromount™, Sigma-Aldrich®
Färbemittel
Mikroskopie- und Färbereagenzien
Fluoromount™ Aqueous Mounting Medium for use with fluorescent dye-stained tissue.
Used for mounting:
- Coverslips on slides for immunofluorescence microscopic examination of PC12 and Hela cells
- Hippocampus neurons cells for confocal microscope studies
- Brain sections for immunofluorescence staining