5-Fluoruracil ≥99% (durch HPLC), Pulver, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr


M-Clarity™ Qualitätsstufe = MQ200

F6627-1G F6627-10G F6627-5G
F6627-1GEA 31.9 EUR
F6627-1G F6627-10G F6627-5G
5-Fluoruracil ≥99% (durch HPLC), Pulver, Sigma-Aldrich®
5-Fluoruracil
Formel: C₄H₃FN₂O₂
Siedepunkt: 347 °C (1013 hPa)
Lagertemperatur: Room Temperature
MDL: MFCD00006018
CAS-Nummer: 51-21-8
EINECS: 200-085-6
UN: 2811
ADR: 6.1,III
Merck Index: 13,04208
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34b30f6a165337be46a329dd264fa290162179a9&vid=0a0335aa0c192ae78b4e943cb4e6a96333d3fb37&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) White to Off-White
Appearance (Form) Powder
Solubility (Colour) Colourless to Very Faint Yellow
Solubility (Turbidity), 50 mg per ml in 1 M Ammonium Hydroxide Clear
Infrared spectrum Conforms to Structure
Purity (HPLC) ≥99%
Recommended Retest Period 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR