Aqueous mounting medium, Fluoroshield™ with PPD, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
F6812-20ML
F6812-20MLEA 97.5 EUR
F6812-20ML
Aqueous mounting medium, Fluoroshield™ with PPD, Sigma-Aldrich®
Färbemittel Mikroskopie- und Färbereagenzien
Fluoroshield™ with PPD (p-phenylenediamine; 1,4-phenylenediamine) is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.
  • Storage temperature: 2 - 8 °C
Fluoroshield prevents rapid photobleaching of FITC, Texas Red®, AMCA, Alexa Fluor® 488, Alexa Fluor 594, and tetramethylrhodamine. This mounting medium is fortified with PPD, an anti-fade reagent used to retard photobleaching of fluorescein and other fluorescent dyes. NOTE: Fluoroshield with PPD is NOT SUITABLE for immunofluorescence techniques involving Cy® dyes, R-phycoerythrin (R-PE), phycocyanin (PC),or allophycocyanin (APC).