Aqueous mounting medium, Fluoroshield™ with 1,4-Diazabicyclo[2.2.2]octane, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
F6937-20ML
F6937-20MLEA
108
EUR
F6937-20ML
Aqueous mounting medium, Fluoroshield™ with 1,4-Diazabicyclo[2.2.2]octane, Sigma-Aldrich®
Färbemittel
Mikroskopie- und Färbereagenzien
Fluoroshield™ with 1,4-Diazabicyclo[2.2.2]octane is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.
- Storage temperature: 2 - 8 °C
Fluoroshield™ prevents rapid photobleaching of FITC, Texas Red®, AMCA, Alexa Fluor™ 488, Alexa Fluor 594, Cy® dyes, and tetramethylrhodamine. This mounting medium is fortified with 1,4-Diazabicyclo[2.2.2]octane (a generic version of DABCO™), an anti-fade reagent used to retard photobleaching of fluorescein and other fluorescent dyes.