Fluoresceindiacetat Zell-Lebensfähigkeits-Farbe, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY


M-Clarity™ Qualitätsstufe = MQ200

F7378-100G F7378-10G F7378-25G F7378-5G
F7378-100GEA 772 EUR
F7378-100G F7378-10G F7378-25G F7378-5G
Fluoresceindiacetat Zell-Lebensfähigkeits-Farbe, Sigma-Aldrich®
Fluoresceindiacetat

Fluorescein diacetate (FDA) is a non-fluorescent molecule, which is hydrolysed to fluorescent fluorescein in live cells. Dead cells cannot accumulate or hydrolyse FDA. Fluorescein diacetate has been used for the differentiation of live cells from dead cells.

Formel: C₂₄H₁₆O₇
Lagertemperatur: Room Temperature
MDL: MFCD00005062
CAS-Nummer: 596-09-8
EINECS: 209-877-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7755e80a40ba77a5c7ea98959a1302a6b8024d53&vid=7755e80a40ba77a5c7ea98959a1302a6b8024d53&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) White to Faint Yellow
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Solubility (Turbidity), c = 25 mg/ml; Acetone Clear
Solubility (Colour) Colourless to Faint Yellow
Emission Wavelength, In 0.1 N Sodium Hydroxide 488.0 - 494.0 nm
EMM ≥86.000
Carbon 68.5 - 69.9 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR