Fluoresceindiacetat Zell-Lebensfähigkeits-Farbe, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
M-Clarity™ Qualitätsstufe = MQ200
Fluorescein diacetate (FDA) is a non-fluorescent molecule, which is hydrolysed to fluorescent fluorescein in live cells. Dead cells cannot accumulate or hydrolyse FDA. Fluorescein diacetate has been used for the differentiation of live cells from dead cells.
Formel:
C₂₄H₁₆O₇ Schmelzpunkt: 200,..203 °C (Lit.) Lagertemperatur: Room Temperature |
MDL:
MFCD00005062 CAS-Nummer: 596-09-8 EINECS: 209-877-6 |
|
Spezifikation Testergebnisse
Appearance (Colour) | White to Faint Yellow |
Appearance (Form) | Powder |
Infrared spectrum | Conforms to Structure |
Solubility (Turbidity), c = 25 mg/ml; Acetone | Clear |
Solubility (Colour) | Colourless to Faint Yellow |
Emission Wavelength, In 0.1 N Sodium Hydroxide | 488.0 - 494.0 nm |
EMM | ≥86.000 |
Carbon | 68.5 - 69.9 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...