Dithiothreitol (DTT, Cleland's Reagenz) 99,0, Biotech Reagent, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyme: DTT, (R*,R*)-1,4-Dimercapto-2,3-butandiol

F781-05EA 18810 EUR
F781-05
Dithiothreitol (DTT, Cleland's Reagenz) 99,0, Biotech Reagent, J.T.Baker®
Dithiothreitol (DTT, Cleland's Reagenz)
Formel: C₄H₁₀O₂S₂
Molecular Weight: 154.25 g/mol
Siedepunkt: ∼125 °C (1013 hPa)
Schmelzpunkt: 42…44 °C
Flammpunkt: >110 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00004877
CAS-Nummer: 3483-12-3
EINECS: 222-468-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product
Keep refrigerated between 2-8°C (36-46°F) and store under nitrogen. ≥99.0%
CAUTION: For Manufacturing, processing or repackaging ≤0.060
Assay (HSCH₂(CHOH)₂CH₂SH) ≥99.0 %
Appearance Passes Test
Absorbance of a 0.1M Solution (1-cm path vs water) at - 280 nm ≤0.060
Absorbance of a 0.1M Solution (1-cm path vs water) at - 260 nm ≤0.400
Infrared Spectrum Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR