Fluorinert™ FC-40, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
M-Clarity™ Qualitätsstufe = MQ200
F9755-250ML
F9755-250MLEA
894
EUR
F9755-250ML
Fluorinert™ FC-40, Sigma-Aldrich®
Fluorinert™
Fluorinert™ FC-40 is an immiscible fluorocarbon oil.
- Suitable for haematology
- Suitable for histology
Fluorinert™ FC-40 has been used to evaluate its compatibility with real time quantitative PCR. It has been used in the cytoskeletal contractility assay to deposit samples in dish.
For possible use as a high density chase solution in density gradient studies.
Bitte beachten Fluorinert is a trademark of 3M Company
Lagertemperatur:
Raumtemperatur |
MDL:
MFCD00131095 CAS-Nummer: 86508-42-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...