FIM-1, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: N-[3-(3-[2,5-Dihydro-4-(1-methyl-1H-indol-3-yl)-2,5-dioxo-1H-pyrrol-3-yl]-1H-indol-1-yl)propyl]-3′,6′-dihydroxy-3-oxospiro[isobenzofuran-1[3H],9′-[9H]xanthene]-5-carboxamide

M-Clarity™ Qualitätsstufe = MQ200

F9926-250UG
F9926-250UGEA 332 EUR
F9926-250UG
FIM-1, Sigma-Aldrich®
FIM-1

A fluorescent conjugate of bisindolylmaleimide that specifically binds phosphokinase C at the active site. Can monitor enzyme activation and translocation. This reagent is not membrane permeable. The diacetate form (Sigma No. F9801) is membrane permeable.

Formel: C₄₅H₃₁N₄O₈
Molecular Weight: 755,75 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD03453075
CAS-Nummer: 220518-50-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR