Protective hand creams, Ligana® A Oleo-Tec
Lieferant: FAWECO HAUTSCHUTZ
12952013
FAWE12952013EA 0 EUR
FAWE12952013
Protective hand creams, Ligana® A Oleo-Tec
Hautpflege Hautcremes Handcremes
Lindesa® O Skin protection cream is especially suitable when working with water-insoluble materials like varnishes, oils, petrol, resins, paints, organic solvents, coating compounds, or similar material, as well as caring for degreased skin after work.
  • Moderately greasy cream
  • Oil in water emulsion
  • Silicone-free, perfumed
Bitte beachten *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904