Hand disinfectant, MYXAL® SEPT 80
Lieferant: FAWECO HAUTSCHUTZ
13661-010
FAWE13661-010EA 0 EUR
FAWE13661-010
Hand disinfectant, MYXAL® SEPT 80
Hautpflege Desinfektionsmittel für die Haut Desinfektionsfluid und -schaum für die Haut
This biocide is skin-friendly due to the combination of active ingredients in a position to achieve a broad spectrum of action with simultaneous skin care by moisturising substances.
  • Used for hygienic hand disinfection
  • Recipe is water-clear, perfume and phenol free
  • Highly flammable liquid and vapour
It is bactericidal (including MRSA), tuberculocidal, yeasticidal, yeasticidal according to European standard and DVV / RKI guideline incl. Noro and Rota viruses and also listed at the Association for Applied Hygiene (VAH).
Can be stored in originalMore Product Information