Restriction enzymes, PstI, FastDigest™, Fermentas
Lieferant: Thermo Fisher Scientific
FD0614
FD0615
FD0614EA
72.25
EUR
FD0614
FD0615
Restriction enzymes, PstI, FastDigest™, Fermentas
Nukleinsäure-Reagenzien
Restriktionsenzym
Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.
- 100% activity of all FastDigest™ enzymes in one universal reaction buffer
- Direct loading on gels with FastDigest™ Green buffer
- 100% buffer compatibility with downstream applications
Designed to complete digestion in 5 to 15 minutes.