Restriction enzymes, SmaI, FastDigest™, Fermentas
Lieferant: Thermo Fisher Scientific
FD0663
FD0663EA
41.41
EUR
FD0663
Restriction enzymes, SmaI, FastDigest™, Fermentas
Nukleinsäure-Reagenzien
Restriktionsenzym
Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.
- 100% activity of all FastDigest™ enzymes in one universal reaction buffer
- Direct loading on gels with FastDigest™ Green buffer
- 100% buffer compatibility with downstream applications
Designed to complete digestion in 5 to 15 minutes.