Restriction enzymes, SpeI, FastDigest™, fermentas
Lieferant: Thermo Fisher Scientific
FD1254
FD1254EA
65.75
EUR
FD1254
Restriction enzymes, SpeI, FastDigest™, fermentas
Nukleinsäure-Reagenzien
Restriktionsenzym
Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.
- 100% activity of all FastDigest™ enzymes in one universal reaction buffer
- Designed to complete digestion in 5 to 15 minutes
- Direct loading on gels with FastDigest™ Green buffer
- 100% buffer compatibility with downstream applications