Autopsy blade handles and blades, FEATHER®
Lieferant: PFM Medical GmbH
205000170 205000100 205000000 205000325
FEAT205000170EA 210 EUR
FEAT205000170 FEAT205000100 FEAT205000000 FEAT205000325
Autopsy blade handles and blades, FEATHER®
Mikrotome und Mikrotomanordnungen Mikrotomklingen und -griffe
The plastic handle is autoclavable and features excellent chemical resistance. It can be easily disassembled for cleaning. Superior sharpness and cutting performance compared to conventional knives. The same handle can be used for brain dissection and other purposes simply by exchanging the blade.
  • Fast and easy blade change
  • Handle is easy to clean and autoclavable