Lidocain

Lieferant: Fitzgerald Industries

10-L40-M24072
FITZ10-L40-M24072EA 0 EUR
FITZ10-L40-M24072
Lidocain
Lidocain
Formel: C₁₄H₂₂N₂O
Molecular Weight: 234.34 g/mol
MDL: MFCD00026733
CAS-Nummer: 137-58-6
EINECS: 205-302-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR