Anti-Theophylline Mouse Monoclonal Antibody

Lieferant: Fitzgerald Industries
10-T10-M94101
FITZ10-T10-M94101EA 0 EUR
FITZ10-T10-M94101
Anti-Theophylline Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Theophylline
Clonality: Monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR