Vancomycin Hydrochlorid

Lieferant: Fitzgerald Industries

10-V10-M997129
FITZ10-V10-M997129EA 0 EUR
FITZ10-V10-M997129
Vancomycin Hydrochlorid
Vancomycin Hydrochlorid
Formel: C₆₆H₇₅Cl₂N₉O₂₄·HCl
MDL: MFCD03613611
CAS-Nummer: 1404-93-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR