Anti-HSA Rabbit Polyclonal Antibody
Lieferant: Fitzgerald Industries
20-AR05
FITZ20-AR05EA 0 EUR
FITZ20-AR05
Anti-HSA Rabbit Polyclonal Antibody
Antikörper
Human serum albumin is the most abundant protein in human blood plasma. It is produced in the liver. Albumin comprises about half of the blood serum protein. It is soluble and monomeric. The gene for albumin is located on chromosome 4 and mutations in this gene can result in various anomalous proteins. The human albumin gene is 16,961 nucleotides long from the putative ?cap? site to the first poly(A) addition site. It is split into 15 exons that are symmetrically placed within the 3 domains thought to have arisen by triplication of a single primordial domain.