G418

Lieferant: Fitzgerald Industries
Achtung

Synonyme: G-418, (2R,3S,4R,5R,6S)-5-Amino-6-(((1R,2S,3S,4R,6S)-4,6-diamino-3-(((2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-2-((R)-1-hydroxyethyl)tetrahydro-2H-pyran-3,4-diol

20-GG15
FITZ20-GG15EA 0 EUR
FITZ20-GG15
G418
G418
Formel: C₂₀H₄₀N₄O₁₀
Molecular Weight: 496.552 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30541267
CAS-Nummer: 49863-47-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR