Progesteron

Lieferant: Fitzgerald Industries

20-PR20
FITZ20-PR20EA 0 EUR
FITZ20-PR20
Progesteron
Progesteron
Formel: C₂₁H₃₀O₂
MDL: MFCD00003658
CAS-Nummer: 57-83-0
EINECS: 200-350-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR