Tobramycin
Lieferant: Fitzgerald Industries
|
Gefahr
|
Synonyme:
(2S,3R,4S,5S,6R)-4-Amino-2-(((1S,2S,3R,4S,6R)-4,6-diamino-3-(((2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxytetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,5-diol
20-TG30
FITZ20-TG30EA
0
EUR
FITZ20-TG30
Tobramycin
Tobramycin
Formel:
C₁₈H₃₇N₅O₉ Molecular Weight: 467,51 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00077885 CAS-Nummer: 32986-56-4 EINECS: 251-322-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...