Tobramycin

Lieferant: Fitzgerald Industries
Gefahr

20-TS30
FITZ20-TS30EA 0 EUR
FITZ20-TS30
Tobramycin
Tobramycin
Formel: C₁₈H₃₇N₅O₉
Lagertemperatur: Kühlschrank
MDL: MFCD00077885
CAS-Nummer: 32986-56-4
EINECS: 251-322-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR