Estriol

Lieferant: Fitzgerald Industries

30-AE31
FITZ30-AE31EA 0 EUR
FITZ30-AE31
Estriol
Estriol
Formel: C₁₈H₂₄O₃
MDL: MFCD00003691
CAS-Nummer: 50-27-1
EINECS: 200-022-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR