Vacuum wand, C-series
Lieferant: WINDRUSH TECHNOLOGY
C003-Y-98-CP 106518 90-CP C002-Y-92-CP C002-Y-90-CP C003-X-95-CP 98-CP C003-Y-99-CP C003-Y-91-CP C002-X-95-CP C003-Y-92-CP C002-Y-98-CP C003-X-96-CP C001-Y-91-CP C002-X-96-CP 99-CP 91-CP C002-Y C003-Y-90-CP
FLUMC003-Y-98-CPEA 0 EUR
FLUMC003-Y-98-CP FLUM106518 FLUM90-CP FLUMC002-Y-92-CP FLUMC002-Y-90-CP FLUMC003-X-95-CP FLUM98-CP FLUMC003-Y-99-CP FLUMC003-Y-91-CP FLUMC002-X-95-CP FLUMC003-Y-92-CP FLUMC002-Y-98-CP FLUMC003-X-96-CP FLUMC001-Y-91-CP FLUMC002-X-96-CP FLUM99-CP FLUM91-CP FLUMC002-Y FLUMC003-Y-90-CP
Vacuum wand, C-series
Vakuum-Systeme
This C series wand bodies are manufactured in ESD safe materials including PEEK tips are the safest means of wafer handling. The tips are optimally sized for each wafer diameter, ensuring that contact is both minimal and secure.
  • Polished contact surface provides excellent adhesion to water
  • Fixed tip and swivel joint body styles are available
  • Unique control valve ensures reliable suction and instantaneous release without particle generation