Vacuum wand, F-series
Lieferant: WINDRUSH TECHNOLOGY
03-3F F003-X-06-VP F003-X-02-VP F003-X-02-3F F001-Z-01-VP F002-X-73-3F F001-Y-14-3F F001-X-01-3F F003-Y-11-3F F001-Y-14-PK F002-Y-11-3F F002-X-03-VP F002-X-06-3F F003-Y-14-3F F003-X-06-3F F003-X-06-PK F003-Z-05-3F F003-Z-05-PK F003-Z-05-VP
FLUM03-3FEA 0 EUR
FLUM03-3F FLUMF003-X-06-VP FLUMF003-X-02-VP FLUMF003-X-02-3F FLUMF001-Z-01-VP FLUMF002-X-73-3F FLUMF001-Y-14-3F FLUMF001-X-01-3F FLUMF003-Y-11-3F FLUMF001-Y-14-PK FLUMF002-Y-11-3F FLUMF002-X-03-VP FLUMF002-X-06-3F FLUMF003-Y-14-3F FLUMF003-X-06-3F FLUMF003-X-06-PK FLUMF003-Z-05-3F FLUMF003-Z-05-PK FLUMF003-Z-05-VP
Vacuum wand, F-series
Vakuum-Systeme
This F series wand bodies are manufactured in teflon and are the cleanest means of wafer handling. Tips are available in PCTFE, PEEK, Vespel and stainless steel. The tips are optimally sized for each wafer diameter, ensuring that contact is both minimal and secure.
  • Polished contact surface provides excellent adhesion to water
  • Fixed tip, collect joint and swivel joint are three body styles
  • Unique control valve ensures reliable suction and instantaneous release with out particle genertion