α-Brom-2-chlortoluol
Lieferant: FLUOROCHEM
Synonyme:
2-Chlorbenzylbromid
001008-25G
001008-100G
FLUO001008-25GEA
42.9
EUR
FLUO001008-25G
FLUO001008-100G
α-Brom-2-chlortoluol
α-Brom-2-chlortoluol
Formel:
C₇H₆BrCl Molecular Weight: 205.48 g/mol Siedepunkt: 235 Dichte: 1.5897 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000566 CAS-Nummer: 611-17-6 EINECS: 210-257-2 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...