N-Butylbenzylamin

Lieferant: FLUOROCHEM

Synonyme: N-Benzyl-n-butylamine

001047-25G
FLUO001047-25GEA 183 EUR
FLUO001047-25G
N-Butylbenzylamin
N-Butylbenzylamin
Formel: C₁₁H₁₇N
MDL: MFCD00009427
CAS-Nummer: 2403-22-7
EINECS: 219-287-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR