2,3-Dibrombutan
Lieferant: FLUOROCHEM
001074-100G
001074-1G
FLUO001074-100GEA
72.9
EUR
FLUO001074-100G
FLUO001074-1G
2,3-Dibrombutan
2,3-Dibromobutane
Formel:
H₃C(BrCH)₂CH₃ Molecular Weight: 215.92 g/mol Siedepunkt: 103…108 °C (160 mmHg) Schmelzpunkt: –24 °C Dichte: 1.83 g/cm³ (0 °C) |
MDL:
MFCD00000146 CAS-Nummer: 5408-86-6 EINECS: 226-476-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...