3-Hydroxy-2,4,6-tribrombenzoesäure

Lieferant: FLUOROCHEM

Synonyme: 2,4,6-Tribromo-3-hydroxybenzoic acid

001084-5G
FLUO001084-5GEA 0 EUR
FLUO001084-5G
3-Hydroxy-2,4,6-tribrombenzoesäure
3-Hydroxy-2,4,6-tribromobenzoic acid
Formel: C₇H₃Br₃O₃
Molecular Weight: 374,81 g/mol
Siedepunkt: 396 °C (1013 hPa)
Schmelzpunkt: 146 °C
MDL: MFCD00055557
CAS-Nummer: 14348-40-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR