Tetramethylensulfoxid

Lieferant: FLUOROCHEM

Synonyme: Tetrahydrothiophen-1-oxid, Thiacyclopentan-1-oxid

001093-25G
FLUO001093-25GEA 198 EUR
FLUO001093-25G
Tetramethylensulfoxid
Tetramethylensulfoxid
Formel: C₄H₈OS
Molecular Weight: 104,17 g/mol
MDL: MFCD00005477
CAS-Nummer: 1600-44-8
EINECS: 216-493-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR