2-Bromzimtsäure
Lieferant: FLUOROCHEM
001107-5G
FLUO001107-5GEA
40.1
EUR
FLUO001107-5G
2-Bromzimtsäure
2-Bromzimtsäure
Formel:
C₉H₇BrO₂ |
MDL:
MFCD00016836 CAS-Nummer: 7499-56-1 EINECS: 231-353-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...