3-Bromocinnamic acid

Lieferant: FLUOROCHEM
Achtung

Synonyme: 3-(3-Bromophenyl)acrylic acid, 3-(3-Bromophenyl)prop-2-enoic acid

001108-25G
FLUO001108-25GEA 58.5 EUR
FLUO001108-25G
3-Bromocinnamic acid
3-Bromocinnamic acid
Formel: C₉H₇BrO₂
Molecular Weight: 227,055 g/mol
MDL: MFCD00004382
CAS-Nummer: 32862-97-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR