1,3-Dichlor-4-iodbenzol
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
2,4-Dichloriodbenzol, 2,4-Dichlor-1-iodbenzol
001167-25G
001167.100G
FLUO001167-25GEA
86
EUR
FLUO001167-25G
FLUO001167.100G
1,3-Dichlor-4-iodbenzol
1,3-Dichlor-4-iodbenzol
Formel:
C₆H₃Cl₂I Molecular Weight: 272,9 g/mol Siedepunkt: 262 °C (1013 hPa) Dichte: 2,01 g/cm³ (20 °C) |
MDL:
MFCD00001035 CAS-Nummer: 29898-32-6 EINECS: 249-938-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...