4-Fluorzimtsäure

Lieferant: FLUOROCHEM

Synonyme: 3-(p-Fluorphenyl)propensäure, 4-Fluorcinnamsäure, 3-(4-Fluorphenyl)propensäure

001180-25G 001180-100G 001180-250G
FLUO001180-25GEA 45.9 EUR
FLUO001180-25G FLUO001180-100G FLUO001180-250G
4-Fluorzimtsäure
4-Fluorzimtsäure
Formel: C₉H₇FO₂
Molecular Weight: 166.15 g/mol
Schmelzpunkt: 209
Dichte: 1 g/cm³ (20 °C)
MDL: MFCD00004395
CAS-Nummer: 459-32-5
EINECS: 207-288-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR